Learn More
Astell Scientific™ Load Support Plate
Product Code. 12795355
Click to view available options
For Use With (Equipment):
AMA250, AMA260, AMA270 Autoclaves
AMB420 and AMB220 Autoclaves
Unit Size:
Each
Product Code. 12795355
Brand: Astell Scientific™ AAN074
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Support shelf to expand autoclave capacity. Astell Scientific™ Load Support Plate accommodates additional Duran bottles without the need for baskets.
  • Expands autoclave loading capacity.
  • Supports additional Duran bottles without the need for baskets.
  • Stainless steel construction.
Specifications
For Use With (Equipment) AMA250, AMA260, AMA270 Autoclaves
Product Type Load Support Plate
Material Stainless Steel
Quantity 1 Ea.