missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
LNX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13124-1-AP-150UL
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
This antibody can detect both LNX1p70 and LNX1p80, around 70-80 kDa and 80-100 kDa, respectively.
Lnx1 (Ligand of Numb Protein X 1) protein is encoded by the Lnx1 gene located on chromosome 4 in humans and is known as PDZRN2 (PDZ domain-containing RING finger 2). It is through its RING finger and PDZ domains that Lnx1 forms oligomers and interacts with some of its target proteins such as JAM4 to form multiprotein complexes. Lnx1 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Phosphotyrosine binding domain (PTB) on Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus positively regulates the Notch signaling pathway.Tekniske data
| LNX1 | |
| Polyclonal | |
| Unconjugated | |
| LNX1 | |
| ligand of numb protein X 1, LNX, LNX1, MPDZ, Numb binding protein 1, PDZRN2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16924, 360926, 84708 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O70263, Q8TBB1 | |
| Lnx1 | |
| LNX1 Fusion Protein Ag3823 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion