missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LNX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | LNX1 |
|---|---|
| Concentration | 0.13 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16801194
|
Proteintech
13124-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16891184
|
Proteintech
13124-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody can detect both LNX1p70 and LNX1p80, around 70-80 kDa and 80-100 kDa, respectively.
Lnx1 (Ligand of Numb Protein X 1) protein is encoded by the Lnx1 gene located on chromosome 4 in humans and is known as PDZRN2 (PDZ domain-containing RING finger 2). It is through its RING finger and PDZ domains that Lnx1 forms oligomers and interacts with some of its target proteins such as JAM4 to form multiprotein complexes. Lnx1 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Phosphotyrosine binding domain (PTB) on Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus positively regulates the Notch signaling pathway.Specifications
| LNX1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O70263, Q8TBB1 | |
| 16924, 360926, 84708 | |
| LNX1 Fusion Protein Ag3823 | |
| Primary | |
| -20°C | |
| LNX1 |
| 0.13 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| ligand of numb protein X 1, LNX, LNX1, MPDZ, Numb binding protein 1, PDZRN2 | |
| Lnx1 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title