Learn More
LNX1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13124-1-AP-20UL
 View more versions of this product
Product Code. 16801194
  • £135.00 / 20µL
Description
This antibody can detect both LNX1p70 and LNX1p80, around 70-80 kDa and 80-100 kDa, respectively.
Lnx1 (Ligand of Numb Protein X 1) protein is encoded by the Lnx1 gene located on chromosome 4 in humans and is known as PDZRN2 (PDZ domain-containing RING finger 2). It is through its RING finger and PDZ domains that Lnx1 forms oligomers and interacts with some of its target proteins such as JAM4 to form multiprotein complexes. Lnx1 is best studied as a E3 ubiquitin ligase that also contains the NPXY motifs which allows binding to the Phosphotyrosine binding domain (PTB) on Numb protein. This interaction has been shown to be responsible for ubiquitination of Numb, which prevents the proteasomal degradation of Notch intracellular domain (NICD) and thus positively regulates the Notch signaling pathway.
Specifications
Specifications
LNX1
Polyclonal
Unconjugated
LNX1
ligand of numb protein X 1, LNX, LNX1, MPDZ, Numb binding protein 1, PDZRN2
Rabbit
Antigen Affinity Chromatography
RUO
16924, 360926, 84708
-20°C
Liquid
Western Blot
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O70263, Q8TBB1
Lnx1
LNX1 Fusion Protein Ag3823
20 μL
Primary
Human, Mouse, Rat