Learn More
Fisherbrand™ LN2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Shop All Fisherbrand ProductsDescription
- Maintains temperature down to -80°C with liquid nitrogen
Specifications
Specifications
| Description | Factory Installed |
| Temperature Range | -80°C |
| For Use With (Equipment) | Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer |