missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
£180.00 - £304.00
Specifications
| Antigen | Livin |
|---|---|
| Dilution | Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18059493
|
Novus Biologicals
NB100-56145SS |
0.025 mL |
£180.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18700514
|
Novus Biologicals
NB100-56145 |
0.05 mL |
£304.00
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Livin Polyclonal specifically detects Livin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| Livin | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Zinc Finger | |
| Neat whole antisera with 0.05% Sodium Azide | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| BIRC7 | |
| IgG | |
| Unpurified | |
| Livin is a short-lived protein that is thought to be degraded by the ubiquitin-proteasome pathway. Thus users may want to do a time course when examining the effects of treatments on Livin expression. Users are encouraged to refer to the literature for additional information; for example, Ma et al. 2006. 2. |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat | |
| Q96CA5 | |
| 79444 | |
| A synthetic peptide corresponding to amino acids 180-230 of human Livin, isoform alpha and isoform beta was used as immunogen (NP_647478). | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 33 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title