missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Livin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
Brand: Novus Biologicals NB100-56145SS
This item is not returnable.
View return policy
Description
Livin Polyclonal specifically detects Livin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Livin | |
| Polyclonal | |
| Western Blot 1:1000-1:2000, Immunohistochemistry 1:1000-1:5000, Immunoprecipitation 1:50-1:200, Immunohistochemistry-Paraffin 1:1000-1:5000 | |
| Q96CA5 | |
| BIRC7 | |
| A synthetic peptide corresponding to amino acids 180-230 of human Livin, isoform alpha and isoform beta was used as immunogen (NP_647478). | |
| Unpurified | |
| RUO | |
| Primary | |
| Livin is a short-lived protein that is thought to be degraded by the ubiquitin-proteasome pathway. Thus users may want to do a time course when examining the effects of treatments on Livin expression. Users are encouraged to refer to the literature for additional information; for example, Ma et al. 2006. 2. | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Neat whole antisera with 0.05% Sodium Azide | |
| baculoviral IAP repeat containing 7, baculoviral IAP repeat-containing 7, KIAPRING finger protein 50, Kidney inhibitor of apoptosis protein, Livin, livin inhibitor-of-apoptosis, Melanoma inhibitor of apoptosis protein, MLIAPlivin inhibitor of apoptosis, ML-IAPmliap, RNF50LIVINbaculoviral IAP repeat-containing protein 7 | |
| Rabbit | |
| 33 kDa | |
| 0.025 mL | |
| Apoptosis, Zinc Finger | |
| 79444 | |
| Human, Mouse, Rat | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction