missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ Violet Viability/Vitality Kit, for 405 nm excitation

Product Code. 10649624
Click to view available options
Quantity:
1 kit
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10649624

Brand: Molecular Probes™ L34958

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

The LIVE/DEAD™ Violet Viability/Vitality Kit provides a two-color fluorescence cell viability and vitality assay that is based on the simultaneous determination of live and dead cells with two probes that measure recognized parameters of cell health.

  • Provides a two-color fluorescence cell viability and vitality assay
  • Based on the simultaneous determination of live and dead cells with two probes that measure recognized parameters of cell health: plasma membrane integrity and intracellular esterase activity
  • Calcein violet AM and aqua-fluorescent reactive dye are optimal dyes for this application
  • Both dyes utilize the violet laser allowing other laser lines to be used for conventional fluorochromes
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Cell Type Eukaryotic Cells
Color Aqua, Violet
Content And Storage Contains 5 vials of calcein violet stain (25 μg/vial), 5 vials of aqua-fluorescent reactive dye, and 1 vial of DMSO.

Store in freezer (-5°C to -30°C) and protect from light.
Description LIVE/DEAD™ Violet Viability/Vitality Kit, for 405 nm excitation
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Stain
Dye Type Other Label(s) or Dye(s)
Emission Aqua-fluorescent reactive dye: 367/526, Calcein violet stain: 400/452
Excitation Wavelength Range 367/400 nm
Form Lyophilized
Format Tube(s)
Product Line LIVE/DEAD
Quantity 1 kit
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.