missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation

Product Code. 17418262
Change view
Click to view available options
Quantity:
200 Assays
400 Assays
80 Assays
Unit Size:
Each
Pack of 200
Pack of 400
Pack of 80
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17418262 200 Assays Pack of 200
17497772 80 Assays Each
17408262 80 Assays Pack of 80
17428262 400 Assays Pack of 400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17418262 Supplier Invitrogen™ Supplier No. L34981
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The LIVE/DEAD Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry.

  • Fixable viability kits allow for clear determination of live and dead cells, enabling more precise analysis with both conventional and spectral flow cytometry
  • Cells labeled with LIVE/DEAD Fixable viability dyes can be washed, fixed, permeabilized, and stained for intracellular antigens while maintaining distinct live and dead cell populations
  • The narrow and unique emission spectra are ideal for expanding high-parameter flow cytometry experiments
  • Designed to provide a bright signal with minimal overlap into adjacent channels, simplifying spectral compensation
  • Since live and dead cells can be discriminated using a single flow cytometer channel, these viability dyes are an ideal choice for multi-color experiments.
  • Their unique spectral fingerprints are a valuable addition to spectral flow cytometry experiments
  • Dyes have been conveniently packaged in single-use vials to help ensure dye stability and performance over time
  • Dissolve the dye in the included anhydrous DMSO and use immediately, or store at -20°C, avoiding freeze-thaw cycles
  • The LIVE/DEAD Fixable Dead Cell Stain Sampler Kit provides one vial (40 assays) of each of eight available dead cell stains
TRUSTED_SUSTAINABILITY

Specifications

Quantity 200 Assays
Description LIVE/DEAD™ Fixable Near IR (876) Viability Kit, for 808 nm excitation
Format Tubes
Color Near-IR
Content And Storage Contains 5 vials of LIVE/DEAD fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Assay Viability Assay
Compatible Cells Eukaryotic Cells
Excitation/Emission 849/876 nm
Flow Cytometer Laser Lines 808 nm
Solubility DMSO (Dimethylsulfoxide)
Shipping Condition Room Temperature
Product Line LIVE/DEAD
For Use With (Equipment) Flow Cytometer
Detection Method Fluorescence
Conjugate LIVE/DEAD Fixable Near IR (876) Cell Stain
For Use With (Application) Flow Cytometry
Product Type Cell Viability Kit
Show More Show Less
What is the concentration of the LIVE/DEAD Fixable dyes?

The concentration of LIVE/DEAD Fixable dyes is proprietary.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.