missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LIVE/DEAD™ Fixable Green Dead Cell Stain Kit, for 488 nm excitation

Product Code. 15507474
Change view
Click to view available options
Quantity:
200 Assays
400 Assays
80 Assays
Unit Size:
1 set
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15507474 80 Assays Each
15678590 400 Assays 1 set
10237152 200 Assays Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15507474 Supplier Invitrogen™ Supplier No. L34969

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

The LIVE/DEAD™ Fixable Green Dead Cell Stain Kit is used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining or prior to elimination of biohazardous materials using formaldehyde fixation
  • Kit has been optimized and validated for use with a blue laser flow cytometer
  • Conveniently packaged in 40-test vials to help ensure the stability and performance of the dye over time
  • Amine reactive dyes in solution will lose their effectiveness over a short period of time, therefore it is recommended to completely use the vial once rehydrated
  • If this is not possible, aliquot the vials in small volumes and store at -80°C, avoiding freeze-thaw cycles
  • LIVE/DEAD™ Fixable Dead Cell Stains are available in a wide variety of colors to meet multi-color panel needs

Optimal brightness

  • The LIVE/DEAD™ Fixable Green stain was selected based on its fluorescent properties to provide a bright signal when excited with a blue 488 nm laser
  • The green-fluorescent reactive dye has an excitation maximum of ∼495 nm making it ideal for use with the blue laser and an emission of ∼520 nm allowing collection in the FITC channel
  • Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, it is an ideal choice for multi-color experiments
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Cell Type Eukaryotic Cells
Color Green
Content And Storage Contains 2 vials of LIVE/DEAD™ fixable dead cell stain and 500 μL DMSO.

Store at -20°C.
Description LIVE/DEAD™ Fixable Green Dead Cell Stain Kit, for 488 nm excitation
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Stain
Dye Type Other Labels or Dyes
Emission 520
Excitation Wavelength Range 495 nm
Form Solid
Format Tube(s)
Product Line LIVE/DEAD
Quantity 80 Assays
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.