missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ LIVE/DEAD™ Fixable Dead Cell Stain Sampler Kit

Product Code. 10637015
Change view
Click to view available options
Quantity:
1 kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10637015 1 kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 10637015 Supplier Molecular Probes™ Supplier No. L34960

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

LIVE/DEAD™ Fixable dead cell stains are used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation.

  • Used to determine the viability of cells prior to the fixation and permeabilization required for intracellular antibody staining, or prior to elimination of biohazardous materials using formaldehyde fixation
  • The stains in the sampler kit have been optimized and validated for use in flow cytometry
  • Stains in this sampler kit have been conveniently packaged in eight 40-test vials to help ensure the stability and performance of the dye over time
  • Amine-reactive dyes in solution will lose their effectiveness over a short period of time
  • It is recommended to use the vial completely once rehydrated or aliquot the solution and store at -80°C, avoiding freeze thaw cycles
  • LIVE/DEAD™ Fixable dead cell stains are available in a wide variety of colors to meet multi-color panel needs
  • The sampler kit provides one vial (40 assays) of each of the eight dead cell stains available

Optimal brightness

  • The LIVE/DEAD™ Fixable dead cell stains were selected based on their fluorescent properties to provide a bright signal when excited with the optimal lasers
  • Since live and dead cells can be discriminated using a single dye and a single channel of a flow cytometer, these dead cell stains are an ideal choice for multi-color experiments

Find the LIVE/DEAD™ Fixable Dead Cell Stain that fits various needs:

  • For UV lasers: blue
  • For violet lasers: aqua, violet, or yellow
  • For blue lasers: green or red
  • For red lasers: far-red or near-IR
TRUSTED_SUSTAINABILITY

Specifications

Cell Permeability Impermeant
Cell Type Eukaryotic Cells
Color Various
Content And Storage Contians 8 vials (1 vial per color) of the LIVE/DEAD™ fixable dead cell stains and 500 μL DMSO.

Store at -20°C.
Description LIVE/DEAD™ Fixable Dead Cell Stain Sampler Kit
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Flow Cytometer
Product Type Dead Cell Stain Sampler Kit
Dye Type Other Label(s) or Dye(s)
Emission 450/451/526/575/520/615/665/775
Excitation Wavelength Range 350/416/367/400/495/595/650/750 nm
Form Solid
Format Tube(s)
Product Line LIVE/DEAD, Molecular Probes
Quantity 1 kit
Shipping Condition Room Temperature
Solubility DMSO (Dimethylsulfoxide)
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.