missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Litmus Paper Roll
Shop All Fisherbrand Products
Click to view available options
Type:
Litmus Paper, Blue
Litmus Paper, Red
Unit Size:
4 units
Specifications
Specifications
| pH Range | 5 to 8 |
| Format | Roll |
| For Use With (Application) | For orientation and adjustment of pH values |
| Type | Litmus Paper, Blue |
| Length (Metric) | 5 m |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction