Learn More
Lithium titanium oxide, Thermo Scientific Chemicals
£53.60 - £222.00
Chemical Identifiers
CAS | 12031-82-2 |
---|---|
Molecular Formula | Li2O3Ti |
Molecular Weight (g/mol) | 109.74 |
MDL Number | MFCD00016181 |
InChI Key | AXQWGBXVDWYWDE-UHFFFAOYSA-N |
Synonym | lithium titanate, lithium titanium oxide, dilithium dioxido oxo titanium, 2li.o3ti, dilithium 1+ ion oxotitaniumbis olate, dilithium 1+ oxotitaniumbis olate |
IUPAC Name | titanium(4+) dilithium(1+) trioxidandiide |
SMILES | [Li+].[Li+].[O--].[O--].[O--].[Ti+4] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11356738
|
Thermo Scientific Alfa Aesar
013411.18 |
50 g |
£53.60
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11366738
|
Thermo Scientific Alfa Aesar
013411.30 |
250 g |
£222.00
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lithium titanium oxide is used in battery, porcelain enamels and ceramic insulating bodies. It is frequently utilized as a flux due to its good stability. It is used as a cathode in layer one of a double layer cathode for molten carbonate fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12031-82-2 | |
109.74 | |
AXQWGBXVDWYWDE-UHFFFAOYSA-N | |
titanium(4+) dilithium(1+) trioxidandiide |
Li2O3Ti | |
MFCD00016181 | |
lithium titanate, lithium titanium oxide, dilithium dioxido oxo titanium, 2li.o3ti, dilithium 1+ ion oxotitaniumbis olate, dilithium 1+ oxotitaniumbis olate | |
[Li+].[Li+].[O--].[O--].[O--].[Ti+4] |
Specifications
1520°C to 1564°C | |
White | |
Li2O3Ti | |
lithium titanate, lithium titanium oxide, dilithium dioxido oxo titanium, 2li.o3ti, dilithium 1+ ion oxotitaniumbis olate, dilithium 1+ oxotitaniumbis olate | |
AXQWGBXVDWYWDE-UHFFFAOYSA-N | |
titanium(4+) dilithium(1+) trioxidandiide | |
109.78 | |
Lithium titanium oxide |
12031-82-2 | |
Powder | |
MFCD00016181 | |
Insoluble in water. | |
[Li+].[Li+].[O--].[O--].[O--].[Ti+4] | |
109.74 | |
Odorless |
Safety and Handling
missing translation for 'einecsNumber' : 234-759-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only