missing translation for 'onlineSavingsMsg'
Learn More

Lithium titanium oxide

Product Code. 11356738
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356738 50 g 50g
11366738 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11356738 Supplier Thermo Scientific Alfa Aesar Supplier No. 013411.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12031-82-2 | Li2O3Ti | 109.74 g/mol

Lithium titanium oxide is used in battery, porcelain enamels and ceramic insulating bodies. It is frequently utilized as a flux due to its good stability. It is used as a cathode in layer one of a double layer cathode for molten carbonate fuel cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-82-2
Molecular Formula Li2O3Ti
Molecular Weight (g/mol) 109.74
MDL Number MFCD00016181
InChI Key AXQWGBXVDWYWDE-UHFFFAOYSA-N
Synonym lithium titanate, lithium titanium oxide, dilithium dioxido oxo titanium, 2li.o3ti, dilithium 1+ ion oxotitaniumbis olate, dilithium 1+ oxotitaniumbis olate
IUPAC Name titanium(4+) dilithium(1+) trioxidandiide
SMILES [Li+].[Li+].[O--].[O--].[O--].[Ti+4]

Specifications

Melting Point 1520°C to 1564°C
Color White
Physical Form Powder
Quantity 50 g
Solubility Information Insoluble in water.
Formula Weight 109.78
Odor Odorless
Chemical Name or Material Lithium titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.