missing translation for 'onlineSavingsMsg'
Learn More

Lithium niobium oxide, Puratronic™, 99.998% (metals basis excluding Ta), Ta <50ppm

Product Code. 11356827
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11356827 500 g 500g
11336827 25 g 25g
11346827 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11356827 Supplier Thermo Scientific Alfa Aesar Supplier No. 010741.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12031-63-9 | LiNbO3 | 147.84 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-63-9
Molecular Formula LiNbO3
Molecular Weight (g/mol) 147.84
MDL Number MFCD00011080
InChI Key GQYHUHYESMUTHG-UHFFFAOYSA-N
Synonym Lithium niobate
PubChem CID 23665650
IUPAC Name lithium;oxido(dioxo)niobium
SMILES [Li+].[O-][Nb](=O)=O

Specifications

Melting Point ∼1275°C
Physical Form Powder
Quantity 500 g
Formula Weight 147.85
Percent Purity 99.998%
Grade Puratronic
Density 4.3 g/mL
Assay (metals basis excluding Ta), Ta <50ppm
Chemical Name or Material Lithium niobium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.