accessibility menu, dialog, popup

UserName

CAS RN 12031-63-9

LiNbOOO

CAS RN 12031-63-9

IUPAC Name: niobium(5+) lithium(1+) trioxidandiide
Synonyms: lithium niobate
Molecular Weight (g/mol): 147.84
Molecular Formula: LiNbO3
InChi Key: PSVBHJWAIYBPRO-UHFFFAOYSA-N
SMILES: [Li+].[O--].[O--].[O--].[Nb+5]
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (3)