missing translation for 'onlineSavingsMsg'
Learn More

Lithium niobium oxide, 99.99%

Product Code. p-7023843
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
This item is not returnable. View return policy

Product Code. 11453398

Brand: Thermo Scientific Alfa Aesar A17805.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 12031-63-9 | LiNbO3 | 147.84 g/mol

It is employed in the manufacture of optical waveguides and surface acoustic wave devices, Lithium niobate (CAS NO.12031-63-9) is used extensively in the telecoms market, e.g. in mobile telephones and optical modulators. It can also find its uses in laser frequency doubling, nonlinear optics, Pockels cells, optical parametric oscillators, Q-switching devices for lasers, other acousto-optic devices, optical switches for gigahertz frequencies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the manufacture of optical waveguides and surface acoustic wave devices, Lithium niobate (CAS NO.12031-63-9) is used extensively in the telecoms market, e.g. in mobile telephones and optical modulators. It can also find its uses in laser frequency doubling, nonlinear optics, Pockels cells, optical parametric oscillators, Q-switching devices for lasers, other acousto-optic devices, optical switches for gigahertz frequencies.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed in a cool, dry and well ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12031-63-9
Molecular Formula LiNbO3
Molecular Weight (g/mol) 147.84
MDL Number MFCD00011080
InChI Key GQYHUHYESMUTHG-UHFFFAOYSA-N
Synonym Lithium niobate
PubChem CID 23665650
SMILES [Li+].[O-][Nb](=O)=O

Specifications

Melting Point ∼1275°C
Quantity 50 g
Solubility Information Soluble in water.
IUPAC Name lithium;oxido(dioxo)niobium
Formula Weight 147.85
Percent Purity 99.99%
Odor Odorless
Density 4.3
Chemical Name or Material Lithium niobium oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.