Learn More
Agilent BioTek Liquid Handling Control Software, USB Delivery
GREENER_CHOICE
Product Code. 17844677 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems
Unit Size:
Each
Product Code. 17844677
Brand: Agilent BioTek LHC3USB
Liquid Handling Control Software provides a powerful yet flexible interface for use with washers, dispensers and combination liquid handling systems. This USB delivery is to be purchased in conjunction with LHC3 or LHC3SECURE.
  • LHC software makes it easy to program, even the most complex protocols with a choice of deployment
  • Protocols can be run directly from the computer or downloaded to the instrument for convenient onboard recall and operation
  • Full instrument control
  • For use with 405 TS Washer, 405 LS Washer, ELx405 Select Deep Well Washer, 50 TS Washer, MultiFlo Dispenser, MultiFlo FX Dispenser, EL406 Washer Dispenser, 406 FX Washer Dispenser
  • Batch processing with connected BioStack Microplate Stacker or BioSpa 8 Automated Incubator
  • Seamless robotic integration to commercially available robotic systems
TRUSTED_SUSTAINABILITY
Specifications
Includes USB Flash Drive Delivery
For Use With (Equipment) Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems
Type Liquid Handling Control Software