Learn More
Agilent BioTek Liquid Handling Control Software
Alle ansehen Agilent BioTek Produkte
missing translation for 'orderingAttributeHoverText'
Includes:
E-delivery, 5 Installation Licenses
E-delivery, 5 Installation Licenses, 21CFR Part 11 Compliance
missing translation for 'unitSize'
Each
Beschreibung
- LHC software makes it easy to program, even the most complex protocols with a choice of deployment
- Protocols can be run directly from the computer or downloaded to the instrument for convenient onboard recall and operation
- Full instrument control
- For use with 405 TS Washer, 405 LS Washer, ELx405 Select Deep Well Washer, 50 TS Washer, MultiFlo Dispenser, MultiFlo FX Dispenser, EL406 Washer Dispenser, 406 FX Washer Dispenser
- Batch processing with connected BioStack Microplate Stacker or BioSpa 8 Automated Incubator
- Seamless robotic integration to commercially available robotic systems
Spezifikation
Spezifikation
| Includes | E-delivery, 5 Installation Licenses, 21CFR Part 11 Compliance |
| For Use With (Equipment) | Agilent BioTek Microplate Washers, Dispensers, and Liquid Handling Systems |
| Type | Liquid Handling Control Software |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur