missing translation for 'onlineSavingsMsg'
Learn More

LIPH Mouse anti-Human, Porcine, Clone: 1F5D10, Proteintech

Mouse Monoclonal Antibody

£135.00 - £349.00

Specifications

Antigen LIPH
Clone 1F5D10
Concentration 2 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16858055
View Documents
Proteintech
66303-1-IG-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16848055
View Documents
Proteintech
66303-1-IG-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.
Specifications

Specifications

LIPH
2 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
AH, Lipase member H, lipase, member H, LIPH, LPD lipase related protein, LPDLR, mPA PLA1, mPA PLA1 alpha, MPAPLA1, Phospholipase A1 member B, PLA1B
LIPH
IgG2a
Protein A
Antibody
1F5D10
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Human, Pig
Q8WWY8
100737989, 200879
LIPH Fusion Protein Ag24009
Primary
-20°C
LIPH
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.