missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIPH Mouse anti-Human, Porcine, Clone: 1F5D10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66303-1-IG-20UL
This item is not returnable.
View return policy
Description
This gene encodes a membrane-bound member of the mammalian triglyceride lipase family. It catalyzes the production of 2-acyl lysophosphatidic acid (LPA), which is a lipid mediator with diverse biological properties that include platelet aggregation, smooth muscle contraction, and stimulation of cell proliferation and motility.Specifications
| LIPH | |
| Monoclonal | |
| 2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WWY8 | |
| LIPH | |
| LIPH Fusion Protein Ag24009 | |
| 20 μL | |
| Primary | |
| Human, Pig | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1F5D10 | |
| Unconjugated | |
| LIPH | |
| AH, Lipase member H, lipase, member H, LIPH, LPD lipase related protein, LPDLR, mPA PLA1, mPA PLA1 alpha, MPAPLA1, Phospholipase A1 member B, PLA1B | |
| Mouse | |
| Protein A | |
| RUO | |
| 100737989, 200879 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction