missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN-28A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
£150.00 - £386.00
Specifications
| Applications | Immunohistochemistry, Immunofluorescence |
|---|---|
| Classification | Polyclonal |
| Form | Antisera |
| Target Species | Human, Mouse |
| Isotype | IgG |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18785233
|
Novus Biologicals
NBP1-49537SS |
£150.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
|
18777673
|
Novus Biologicals
NBP1-49537 |
0.1 mL |
£386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIN-28A Polyclonal specifically detects LIN-28A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| Immunohistochemistry, Immunofluorescence | |
| Antisera | |
| IgG |
| Polyclonal | |
| Human, Mouse | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title