missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN-28A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NBP1-49537
This item is not returnable.
View return policy
Description
LIN-28A Polyclonal specifically detects LIN-28A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| LIN-28A | |
| Polyclonal | |
| Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 10-15 ug/ml, Immunohistochemistry-Paraffin 1:1000 | |
| Q9H9Z2, Q8K3Y3 | |
| LIN28A | |
| Partial recombinant human Lin28 protein expressed in E. coli. [Swiss-Prot# Q9H9Z2] | |
| 0.1 mL | |
| Embryonic Stem Cell Markers, Epigenetics, microRNA, Neuroscience, Stem Cell Markers, Stem Cells | |
| 79727 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Whole antisera with 0.1% Sodium Azide | |
| CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1 | |
| Rabbit | |
| Unpurified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Antisera |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction