missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN-28A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£317.00 - £530.00
Specifications
| Antigen | LIN-28A |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18605284
|
Novus Biologicals
NBP3-21298-25ul |
25 μg |
£317.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654604
|
Novus Biologicals
NBP3-21298-100ul |
100 μg |
£530.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIN-28A Polyclonal antibody specifically detects LIN-28A in Human samples. It is validated for ImmunofluorescenceSpecifications
| LIN-28A | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Epigenetics, Neuroscience, Stem Cell Markers, Stem Cells | |
| PBS, pH 7.2, 40% glycerol | |
| 79727 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CSDD1lin-28A, FLJ12457, LIN-28, lin-28 homolog (C. elegans), lin-28 homolog A (C. elegans), Lin-28A, LIN28RNA-binding protein LIN-28, ZCCHC1protein lin-28 homolog A, Zinc finger CCHC domain-containing protein 1, zinc finger, CCHC domain containing 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: KCHFCQSISHMVASCPLKAQQGPSAQGKPTYFREEEEEIHSPTLLPEAQN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title