missing translation for 'onlineSavingsMsg'
Learn More

LIMS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen LIMS1
Concentration 0.21 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16890335
View Documents
Proteintech
20772-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16880335
View Documents
Proteintech
20772-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pinch, also designated particularly interesting new Cys-His protein or NY-REN-48, is a focal adhesion protein that is a component of the ILK-PINCH complex. This complex is a major part of the growth factor and integrin signaling pathway. Pinch is involved in cell differentiation, proliferation and survival by acting as an effector of integrin and growth factor signaling. It is a cytoplasmic protein expressed in most tissues and consists of five LIM domains, a nuclear localization signal and a nuclear export signal. The PINCH-1/ILK complex is regulated by a PINCH-1 related protein PINCH-2, which also forms a complex with ILK.
Specifications

Specifications

LIMS1
Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
Unconjugated
Rabbit
Rat, Mouse, Human
P48059, Q99JW4
110829, 3987, 499443
LIMS1 Fusion Protein Ag14764
Primary
-20°C
LIMS1
0.21 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
LIMS1, PINCH, PINCH 1, PINCH1
LIMS1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.