Learn More
LIMS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20772-1-AP-20UL
 View more versions of this product
Product Code. 16890335
  • £135.00 / 20µL
Description
Pinch, also designated particularly interesting new Cys-His protein or NY-REN-48, is a focal adhesion protein that is a component of the ILK-PINCH complex. This complex is a major part of the growth factor and integrin signaling pathway. Pinch is involved in cell differentiation, proliferation and survival by acting as an effector of integrin and growth factor signaling. It is a cytoplasmic protein expressed in most tissues and consists of five LIM domains, a nuclear localization signal and a nuclear export signal. The PINCH-1/ILK complex is regulated by a PINCH-1 related protein PINCH-2, which also forms a complex with ILK.
Specifications
Specifications
LIMS1
Polyclonal
Unconjugated
LIMS1
LIMS1, PINCH, PINCH 1, PINCH1
Rabbit
Antigen Affinity Chromatography
RUO
110829, 3987, 499443
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P48059, Q99JW4
LIMS1
LIMS1 Fusion Protein Ag14764
20 μL
Primary
Rat, Mouse, Human