missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LIMK1 Recombinant Rabbit Monoclonal Antibody (23GB6425), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | LIMK1 |
|---|---|
| Clone | 23GB6425 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088789
![]() |
Invitrogen™
MA553046 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088128
|
Invitrogen™
MA553047 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIMK1 Recombinant Monoclonal antibody specifically detects LIMK1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| LIMK1 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| P53667, P53668 | |
| 16885, 3984 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6425 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| EC 2.7.11.1; kinase LIMK1; KIZ-1; LIM domain kinase 1; lim kinase 1; LIM motif-containing protein kinase; LIM motif-containing protein kinase 1; LIM-domain containing protein kinase 1; LIM-domain containing, protein kinase; LIM-domain containing, protein kinase 1; Limk; LIMK1; LIMK-1 | |
| A synthesized peptide derived from human LIM Kinase 1 (1-57AA). | |
| Primary | |
| -20°C | |
| LIMK1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title