Learn More
Invitrogen™ LIMK1 Recombinant Rabbit Monoclonal Antibody (23GB6425), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553046
 View more versions of this product
Product Code. 30088789
  • £387.00 / 100µL
Description
LIMK1 is a serine/threonine kinase involved in regulation of actin cytoskeletal changes via phosphorylation of cofilin. LIMK1 has also been implicated in brain development. LIM domains are highly conserved cysteine-rich structures containing 2 zinc fingers. Although zinc fingers usually function by binding to DNA or RNA, the LIM motif probably mediates protein-protein interactions. LIM kinase-1 and LIM kinase-2 belong to a small subfamily with a unique combination of 2 N-terminal LIM motifs and a C-terminal protein kinase domain. LIMK1 is likely to be a component of an intracellular signaling pathway and may be involved in brain development. LIMK1 hemizygosity is implicated in the impaired visuospatial constructive cognition of Williams syndrome.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LIMK1
Recombinant Monoclonal
Unconjugated
LIMK1
EC 2.7.11.1; kinase LIMK1; KIZ-1; LIM domain kinase 1; lim kinase 1; LIM motif-containing protein kinase; LIM motif-containing protein kinase 1; LIM-domain containing protein kinase 1; LIM-domain containing, protein kinase; LIM-domain containing, protein kinase 1; Limk; LIMK1; LIMK-1
Rabbit
Affinity chromatography
RUO
16885, 3984
-20°C
Liquid
Western Blot, Immunocytochemistry
23GB6425
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P53667, P53668
LIMK1
A synthesized peptide derived from human LIM Kinase 1 (1-57AA).
100 μL
Primary
Human, Mouse