missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMD1 Antibody (3F2/G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£150.00 - £315.00
Specifications
| Antigen | LIMD1 |
|---|---|
| Clone | 3F2/G6 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18663859
|
Novus Biologicals
NBP2-50476-0.025ml |
0.025 mL |
£150.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18620937
|
Novus Biologicals
NBP2-50476 |
0.1 mL |
£315.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIMD1 Monoclonal antibody specifically detects LIMD1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
| LIMD1 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Epigenetics | |
| PBS | |
| 8994 | |
| IgG1 | |
| Protein A purified |
| 3F2/G6 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| LIM domain-containing protein 1, LIM domains containing 1 | |
| Recombinant LIMD1 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title