missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMD1 Antibody (3F2/G6) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50476
This item is not returnable.
View return policy
Description
LIMD1 Monoclonal antibody specifically detects LIMD1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| LIMD1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
| LIM domain-containing protein 1, LIM domains containing 1 | |
| Recombinant LIMD1 | |
| 0.1 mL | |
| Epigenetics | |
| 8994 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
| 3F2/G6 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction