missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1/2, p Thr508 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£173.00 - £568.00
Specifications
| Concentration | 1 mg/mL |
|---|---|
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Target Species | Human, Mouse, Rat |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18229290
|
Novus Biologicals
NBP2-54645 |
0.1 mg |
£568.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614319
|
Novus Biologicals
NBP2-54645-0.025mg |
0.025 mg |
£173.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18299671
|
Novus Biologicals
NBP2-54645-0.05MG |
0.05 mg |
£366.00
0.05mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIM Kinase 1/2 Polyclonal specifically detects LIM Kinase 1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.Specifications
| 1 mg/mL | |
| Unconjugated | |
| Human, Mouse, Rat | |
| 3984 | |
| A peptide sequence around phosphorylation site of threonine 508 (R-Y-T (p) -V-V) derived from Human LIMK1 | |
| Primary |
| Polyclonal | |
| Rabbit | |
| EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
| LIMK1 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title