missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIM Kinase 1/2, p Thr508 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-54645-0.05MG
This item is not returnable.
View return policy
Description
LIM Kinase 1/2 Polyclonal specifically detects LIM Kinase 1/2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| LIM Kinase 1/2 | |
| Polyclonal | |
| Unconjugated | |
| Antibody supplied in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150 mM NaCl, and 50% glycerol. with 0.02% sodium azide | |
| LIMK1 | |
| A peptide sequence around phosphorylation site of threonine 508 (R-Y-T (p) -V-V) derived from Human LIMK1 | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| 1 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:10-1:500, Immunofluorescence 1:100-1:200 | |
| EC 2.7.11.1, LIM domain kinase 1, LIM motif-containing protein kinase, LIMKLIMK-1 | |
| Rabbit | |
| 0.05 mg | |
| 3984 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction