Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Alexa Fluor 594 Antibody Labeling Kit
Product Code. 18093774 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
Product Code. 18093774
Brand: Novus Biologicals™ 3350015
Antibody labeling kit
Lightning-Link™ Rapid Antibody Labeling kits enable direct labeling of antibodies, proteins, peptides or other biomolecules and allow conjugations to be set up in seconds, and used within minutes (less than 20 minutes).
Kit Contents:
  • Lightning-Link™ Rapid mix
  • 1 vial of LL Rapid Quencher reagent
  • 1 vial of LL Rapid Modifier reagent
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 mg
Product Type Antibody Labeling Kit
Content And Storage -20°C