Learn More
Novus Biologicals™ Lightning-Link (R) Fluorescein Antibody Labeling Kit
Shop All Bio Techne Products
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Description
Lightning-Link™ antibody labeling kits enable the direct labeling of antibodies, proteins, peptides or other biomolecules for use in R&D applications, drug discovery and the development of diagnostic kits.
Kit Contents:
- 1 vial of LL-modifier reagent
- 1 vial of LL-quencher FD reagent
- 1 glass vial of Lightning-Link™ mix
Specifications
Specifications
| Quantity | 100 μg |
| Product Type | Antibody Labeling Kit |
| Content And Storage | -20°C |