missing translation for 'onlineSavingsMsg'
Learn More

Berthold Technologies LightCompass™ Upgrade From Plus to Professional

Product Code. 31777379 Shop All Berthold Technologies Products
Change view
Click to view available options
Product Type:
Software Upgrade Package
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Unit Size
31777379 Software Upgrade Package Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 31777379 Supplier Berthold Technologies Supplier No. 37854407
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Upgrade to advanced scientific software features for data acquisition, analysis, and reporting with flexible protocols and audit trail support.

Workflow

  • Create or open plate layouts
  • Use preloaded protocols (instrument settings and analysis and export/print settings included)
  • Create new protocols with the protocol wizard or build fully custom protocols
  • Analyze data within the protocol
  • Print results or export to Excel or LIMS

Measurement Modes

  • Reagent dispensing and incubation steps
  • Endpoint
  • Kinetics (including long-term/repeated measurements)
  • Wavelength scan
  • Area scan
  • Multiple measurement modes within one protocol

Data Analysis

  • Standard curves with curve fitting (linear, polynomial, 4PL, smoothed cubic spline, others)
  • Threshold-based classification for qualitative assays
  • Custom formulas
  • Quantitative and qualitative analysis

Data Integrity and Compliance

  • User management with role-based access
  • QC/validation with pass/fail criteria
  • Secure data storage (local file system or remote SQL server)
  • Audit trail of user and data actions
  • FDA 21 CFR Part 11 and EU GMP Annex 11 compliance

Applications

  • ELISA
  • Nucleic acid and protein quantification
  • Reporter gene assays
  • Dose-response assays
  • Kinetic assays
  • BRET
  • TR-FRET and HTRF™

Additional Features in Professional Edition

Includes all Plus features, plus:

  • Unlimited calculation matrices
  • Dilution factor calculation
  • User accounts with configurable levels and password protection
  • Active Directory integration
  • Configurable user hierarchies
  • Audit trail with automated log-file generation
  • Plate Organiser (sample ID definition and preparation)
  • Automated filename generation
  • Control history for validation
  • Assay lot number verification
  • Parameter file selection by barcode

Specifications

Product Type Software Upgrade Package
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.