Learn More
Haldenwanger™ Lids for Porcelain Crucibles
Use these lids with porcelain crucibles. Haldenwanger™ Lids for Porcelain Crucibles are constructed from optimized laboratory porcelain.
Brand:  Haldenwanger™ L 79D/5
Code : 02
Additional Details : Weight : 0.18800kg
 View more versions of this product
Product Code. 12366507
  • £107.06 / Pack of 10
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • Available in a variety of sizes
  • For use with Haldenwanger porcelain crucibles
  • Complies with DIN VDE 0335, type C 100
Specifications
Specifications
Porcelain
52mm
Crucible Lid