missing translation for 'onlineSavingsMsg'
Learn More

Lewis X Trisaccharide, TRC

Product Code. 30012439
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012439 1 mg 1mg
30004884 5 mg 5mg
30003240 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012439 Supplier Toronto Research Chemicals Supplier No. TRCL3950001MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71208-06-5
Molecular Formula C20 H35 N O15
Molecular Weight (g/mol) 529.49
IUPAC Name N-[(3R,4R,5S,6R)-2-hydroxy-6-(hydroxymethyl)-5-[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-4-[(2S,3S,4R,5S,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxyoxan-3-yl]acetamide
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](NC(=O)C)C(O)O[C@H](CO)[C@H]2O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)[C@@H](O)[C@H](O)[C@@H]1O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C20H35NO15/c1-5-10(25)12(27)14(29)19(32-5)36-17-9(21-6(2)24)18(31)33-8(4-23)16(17)35-20-15(30)13(28)11(26)7(3-22)34-20/h5,7-20,22-23,25-31H,3-4H2,1-2H3,(H,21,24)/t5-,7+,8+,9+,10+,11-,12+,13-,14-,15+,16+,17+,18?,19-,20-/m0/s1
Formula Weight 529.2007
Percent Purity >95%
Chemical Name or Material Lewis X Trisaccharide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.