missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene B4 R1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP2-27422SS
This item is not returnable.
View return policy
Description
Leukotriene B4 R1 Polyclonal specifically detects Leukotriene B4 R1 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Leukotriene B4 R1 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and white-cheeked gibbon, 94% in common chimpanzee, 83% in orangutan, and 78% in cow, new world monkey, and rhesus monkey. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 3-5ug/ml∼, Flow Cytometry reported in scientific literature (PMID 28763622), Immunohistochemistry, Immunohistochemistry-Paraffin 10ug/ml∼ | |
| Q15722 | |
| LTB4R | |
| A portion of amino acids 1-60 from human BLT1 was used as the immunogen for this antibody (1st extracellular domain). | |
| 0.025 mg | |
| GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 1241 | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction