missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Leukotriene B4 R1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£177.00 - £319.00
Specifications
| Antigen | Leukotriene B4 R1 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 3-5ug/ml∼, Flow Cytometry reported in scientific literature (PMID 28763622), Immunohistochemistry, Immunohistochemistry-Paraffin 10ug/ml∼ |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18114065
|
Novus Biologicals
NBP2-27422SS |
0.025 mg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18179123
|
Novus Biologicals
NBP2-27422 |
0.1 mg |
£319.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Leukotriene B4 R1 Polyclonal specifically detects Leukotriene B4 R1 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Leukotriene B4 R1 | |
| Western Blot 3-5ug/ml∼, Flow Cytometry reported in scientific literature (PMID 28763622), Immunohistochemistry, Immunohistochemistry-Paraffin 10ug/ml∼ | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Primate | |
| Q15722 | |
| 1241 | |
| A portion of amino acids 1-60 from human BLT1 was used as the immunogen for this antibody (1st extracellular domain). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 0.5 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| chemokine receptor-like 1, CMKRL1, leukotriene B4 receptor, LTBR1, P2Y purinoceptor 7, purinergic receptor P2Y, G-protein coupled, 7 | |
| LTB4R | |
| IgG | |
| Protein A purified | |
| The amino acid sequence used as immunogen for this antibody is 100% homologous in human, and white-cheeked gibbon, 94% in common chimpanzee, 83% in orangutan, and 78% in cow, new world monkey, and rhesus monkey. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title