missing translation for 'onlineSavingsMsg'
Learn More

Leuco Gentian Violet-d6 (Major), TRC

Product Code. 30133668
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133668 50 mg 50mg
17958399 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30133668 Supplier Toronto Research Chemicals Supplier No. TRCL33020350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1173023-92-1
Molecular Formula C25 2H6 H25 N3
Molecular Weight (g/mol) 379.57
Synonym Leucocrystal Violet D6, Benzenamine, 4,4'-[[4-[di(methyl-d3)amino]phenyl]methylene]bis[N,N-dimethyl-
IUPAC Name 4-[[4-[bis(trideuteriomethyl)amino]phenyl]-[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline
SMILES [2H]C([2H])([2H])N(c1ccc(cc1)C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C)C([2H])([2H])[2H]

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C25H31N3/c1-26(2)22-13-7-19(8-14-22)25(20-9-15-23(16-10-20)27(3)4)21-11-17-24(18-12-21)28(5)6/h7-18,25H,1-6H3/i1D3,2D3
Formula Weight 379.2895
Chemical Name or Material Leucocrystal violet D6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.