CAS RN 1173023-92-1
CAS RN 1173023-92-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Leucocrystal Violet D6 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality
2
Leuco Gentian Violet-d6 (Major), TRC
CAS: 1173023-92-1 Molecular Formula: C25 2H6 H25 N3 Molecular Weight (g/mol): 379.57 Synonym: Leucocrystal Violet D6,Benzenamine, 4,4'-[[4-[di(methyl-d3)amino]phenyl]methylene]bis[N,N-dimethyl- IUPAC Name: 4-[[4-[bis(trideuteriomethyl)amino]phenyl]-[4-(dimethylamino)phenyl]methyl]-N,N-dimethylaniline SMILES: [2H]C([2H])([2H])N(c1ccc(cc1)C(c2ccc(cc2)N(C)C)c3ccc(cc3)N(C)C)C([2H])([2H])[2H]