missing translation for 'onlineSavingsMsg'
Learn More

Lercanidipine 5-Desmethyl-5-Propyl Ester, TRC

Product Code. 30009423
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30009423 100 mg 100mg
30004245 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009423 Supplier Toronto Research Chemicals Supplier No. TRCL179025100MG
 Restricted Item
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1797124-83-4
Molecular Formula C38 H45 N3 O6
Molecular Weight (g/mol) 639.7804
Synonym Propyl 1,1,N-Trimethyl-N-(3,3-diphenylpropyl)-2-aminoethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-[(3,3-Diphenylpropyl)methylamino]-1,1-dimethylethyl Propyl Ester, Lercanidipine Impurity C
IUPAC Name 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-propyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)(C)CN(C)CCC(c3ccccc3)c4ccccc4)C

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C38H45N3O6/c1-7-23-46-36(42)33-26(2)39-27(3)34(35(33)30-19-14-20-31(24-30)41(44)45)37(43)47-38(4,5)25-40(6)22-21-32(28-15-10-8-11-16-28)29-17-12-9-13-18-29/h8-20,24,32,35,39H,7,21-23,25H2,1-6H3
Formula Weight 639.3308
Chemical Name or Material Lercanidipine 5- Desmethyl-5-Propyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.