CAS RN 1797124-83-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Lercanidipine 5-Desmethyl-5-Propyl Ester, TRC
CAS: 1797124-83-4 Molecular Formula: C38 H45 N3 O6 Molecular Weight (g/mol): 639.7804 Synonym: Propyl 1,1,N-Trimethyl-N-(3,3-diphenylpropyl)-2-aminoethyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)pyridine-3,5-dicarboxylate,1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic Acid 2-[(3,3-Diphenylpropyl)methylamino]-1,1-dimethylethyl Propyl Ester,Lercanidipine Impurity C IUPAC Name: 5-O-[1-[3,3-diphenylpropyl(methyl)amino]-2-methylpropan-2-yl] 3-O-propyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate SMILES: CCCOC(=O)C1=C(C)NC(=C(C1c2cccc(c2)[N+](=O)[O-])C(=O)OC(C)(C)CN(C)CCC(c3ccccc3)c4ccccc4)C