missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Legionella MWY Selective Agar

Product Code. 10482513
Change view
Click to view available options
Description:
Legionella MWY Selective Agar
Quantity:
10/Pk.
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Quantity unitSize
10482513 Legionella MWY Selective Agar 10/Pk. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10482513 Supplier Thermo Scientific™ Supplier No. PO5071A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Isolate Legionellaceae with this alternative to GVPC media which includes additional selective agents to provide better selectivity and improved identification.

Isolate Legionella using Thermo Scientific™ Legionella MWY Selective Agar, a selective medium that is an alternative to GVPC and conforms to ISO. Achieve optimal recovery of Legionellaceae in short incubation periods from both environmental samples and clinical specimens.

MWY Medium has been successfully tested in the examination of clinical specimens with Bromcresol purple and Bromothymol blue coloring the colonies and aiding in the identification of organisms.

Legionella are aerobic, motile, nutritionally fastidious, pleomorphic, gram-negative rods that require special growth factors in culture media to grow. Transmission occurs through aerosolization or aspiration of contaminated water, not person-to-person. Legionellosis refers to two distinct clinical syndromes: Legionnaires’ disease, which most often presents as severe pneumonia, and Pontiac fever, which is an acute, febrile, self-limiting, viral-like illness. Over forty species of Legionella have been characterized; however, Legionella pneumophila remains the most prevalent etiological agent of Legionnaires’ disease. Legionella can be isolated from respiratory tract secretions of all types, sterile body fluids such as blood, and biopsy material.
  • Tested in accordance with the methods described in ISO 7704:2023

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY

Specifications

Color Traffic black to Jet black, opaque
Content And Storage 6°C to 12°C
Description Legionella MWY Selective Agar
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 17g ± 5%
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.