missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Legionella BYCE Medium without Cysteine

Product Code. 10391783
This item is not returnable. View return policy
Product Code. 10391783 Supplier Thermo Scientific™ Supplier No. PO5028A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Presumptively confirm the presence of Legionellaceae from clinical and environmental samples with this ready-to-use medium.

Presumptively confirm the presence of Legionellaceae from clinical and environmental samples with ready-to-use Thermo Scientific™ Oxoid™ Legionella BCYE Medium without Cysteine. Presumptive confirmation of Legionella is based on the fact that Legionellaceae have an absolute nutritional requirement for L-cysteine. Comparing presence and absence of growth of a sample on Legionella BYCE medium with and without Cysteine can be regarded as providing presumptive confirmation of Legionella species.

Use Legionella BCYE Medium without Cysteine for the presumptive confirmation of Legionella species.

Legionellaceae have an absolute nutritional requirement for L-cysteine. Presumptive Legionella species are subcultured on both Legionella BYCE Medium with Cysteine and Legionella BYCE Medium without Cysteine. After incubation at 35oC, the colonies which have grown on Legionella BYCE Medium with Cysteine, but not on Legionella BYCE Medium without Cysteine, can be regarded as presumptive Legionella spp.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Straw
Content And Storage 2°C to 12°C
Format 90mm Monoplate
Manufacturing Origin Germany
Product Type Agar Plate
Fill Weight 19.5g ± 1.0g
Quantity 10/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.