missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lefty1/Lefty2 Recombinant Rabbit Monoclonal Antibody (23GB6650), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | Lefty1/Lefty2 |
|---|---|
| Clone | 23GB6650 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089947
![]() |
Invitrogen™
MA553088 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089649
|
Invitrogen™
MA553089 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lefty1/Lefty2 Recombinant Monoclonal antibody specifically detects Lefty1/Lefty2 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Lefty1/Lefty2 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| O00292, O75610, P57785, Q64280 | |
| 10637, 13590, 289316, 320202, 498299, 7044 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6650 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 6030463A22Rik; AI450052; AV214969; Ebaf; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; left right determination factor 1; Lefta; LEFTB; left-right determination factor 1; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; left-right determination, factor B; lefty; Lefty protein; Lefty1; lefty-1; LEFTY2; LEFTYA; LEFTYB; Protein lefty-1; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; Tgfb4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317 | |
| A synthesized peptide derived from human LEFTY1+LEFTY2 (330-366AA). | |
| Primary | |
| -20°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title