missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lefty1/Lefty2 Recombinant Rabbit Monoclonal Antibody (23GB6650), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553088
This item is not returnable.
View return policy
Description
Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
Specifications
| Lefty1/Lefty2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| LEFTY1 | |
| 6030463A22Rik; AI450052; AV214969; Ebaf; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; left right determination factor 1; Lefta; LEFTB; left-right determination factor 1; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; left-right determination, factor B; lefty; Lefty protein; Lefty1; lefty-1; LEFTY2; LEFTYA; LEFTYB; Protein lefty-1; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; RGD1561867; Stimulated by retinoic acid gene 3 protein; Stra3; Tgfb4; TGF-beta-4; Transforming growth factor beta-4; UNQ278/PRO317 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 10637, 13590, 289316, 320202, 498299, 7044 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 23GB6650 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O00292, O75610, P57785, Q64280 | |
| LEFTY1, LEFTY2 | |
| A synthesized peptide derived from human LEFTY1+LEFTY2 (330-366AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction