missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lefty-A Antibody (10G2), Novus Biologicals™
Rabbit Monoclonal Antibody
£260.00 - £406.00
Specifications
| Antigen | Lefty-A |
|---|---|
| Clone | 10G2 |
| Dilution | Western Blot 1:500-1:2000, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30030684
|
Novus Biologicals
NBP3-26170-50ul |
50 μL |
£260.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039579
|
Novus Biologicals
NBP3-26170-100ul |
100 μL |
£406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lefty-A Monoclonal antibody specifically detects Lefty-A in Human samples. It is validated for Western Blot, ELISASpecifications
| Lefty-A | |
| Western Blot 1:500-1:2000, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| 7044 | |
| IgG | |
| Affinity purified |
| 10G2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EBAFTGF-beta-4, endometrial bleeding associated factor, Endometrial bleeding-associated factor, LEFTAProtein lefty-A, left-right determination factor 2, Left-right determination factor A, LEFTYATransforming growth factor beta-4, MGC46222, Protein lefty-2, TGFB4endometrial bleeding associated factor (left-right determination, factor A,transforming growth factor beta superfamily) | |
| A synthesized peptide derived from Human Lefty-A [UniProt O00292] | |
| Primary | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title