missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lefty-A Antibody (10G2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26170-50ul
This item is not returnable.
View return policy
Description
Lefty-A Monoclonal antibody specifically detects Lefty-A in Human samples. It is validated for Western Blot, ELISA
Specifications
| Lefty-A | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 7044 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 10G2 | |
| Western Blot 1:500-1:2000, ELISA | |
| EBAFTGF-beta-4, endometrial bleeding associated factor, Endometrial bleeding-associated factor, LEFTAProtein lefty-A, left-right determination factor 2, Left-right determination factor A, LEFTYATransforming growth factor beta-4, MGC46222, Protein lefty-2, TGFB4endometrial bleeding associated factor (left-right determination, factor A,transforming growth factor beta superfamily) | |
| A synthesized peptide derived from Human Lefty-A [UniProt O00292] | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction