missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LECT2 Antibody (MM0449-4B24) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£153.00 - £426.00
Specifications
| Antigen | LECT2 |
|---|---|
| Clone | MM0449-4B24 |
| Concentration | LYOPH |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18047686
|
Novus Biologicals
NBP2-11768-0.025MG |
0.025 mg |
£153.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095637
|
Novus Biologicals
NBP2-11768 |
0.1 mg |
£426.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LECT2 Monoclonal specifically detects LECT2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| LECT2 | |
| LYOPH | |
| Monoclonal | |
| Purified | |
| RUO | |
| O14960 | |
| 3950 | |
| IgG1 | |
| Protein G purified |
| MM0449-4B24 | |
| Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| chm2, chm-II, chondromodulin-II, hLECT2, LECT-2, leukocyte cell-derived chemotaxin 2, leukocyte cell-derived chemotaxin-2, MGC126628 | |
| LECT2 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title